Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, 0.5% In 50% Ethanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™

Manufacturer:  LabChem, Inc. LC182001

 View more versions of this product

Catalog No. LC182001


Add to cart

Description

Description

Specifications

Specifications

Phenolphthalein
49.8, 44.8, 2.5, 2.4, 0.5
C20H14O4
318.32
Certified
Soluble in water
Carbon monoxide; Carbon dioxide
Colorless to Pink
500mL
0.5% In 50% Ethanol
64-17-5, 67-63-0, 67-56-1, 77-09-8
Passes Test
Liquid
Poly Bottle
0.95g/mL
0.95g/mL
8.3 to 10.0
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May cause cancer.
Suspected of damaging the unborn child (oral).
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
Keep container tightly closed.
Ground/bond

Danger

Recommended Storage : Room Temperature

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit