Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phenolphthalein, 1% w/v in alcohol

Manufacturer:  Alfa Aesar™ 38704AP

Quantity 500mL

 View more versions of this product

Catalog No. AA38704AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

77-09-8
MFCD00005913
Phenolphthalein
318.33
UN1987

Specifications

1% w/v in alcohol
77-09-8
Liquid
MFCD00005913
Colorless
Phenolphthalein
500mL
Alcohol-like
318.33
UN1987
11° (52°F)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H350-H341-H361-H371
Highly flammable liquid and vapor.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit