Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5391249
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
77-09-8
C20H14O4
318.33
MFCD00005913
KJFMBFZCATUALV-UHFFFAOYSA-N
phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
4764
CHEBI:34914
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
77-09-8
C20H14O4
318.33
MFCD00005913
KJFMBFZCATUALV-UHFFFAOYSA-N
phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
4764
CHEBI:34914
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

High quality chemicals for laboratory and specialized industrial use

Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 1.30kg
Formula Weight 318.33
Grade BAKER ANALYZED™ ACS Reagent
Physical Form Powder
Chemical Name or Material Phenolphthalein
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit