Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, Reagent ACS Meets USP and ACS Specifications, Reagents

Catalog No. p-200021156
Click to view available options
Quantity:
25 g
100 g
500 g
3 kg
25 lb.
25 kg

Phenolphthalein, Reagent ACS Meets USP and ACS Specifications, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 258°C to 262°C
Boiling Point 558°C
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.299
pH Color Changes Colorless (pH 8.0) to Pink (pH 10.0)
Color White to Yellow
Quantity 100 g
Physical Form Solid
Chemical Name or Material Phenolphthalein, Reagent ACS Meets USP and ACS Specifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.