Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenosafranin, Acros Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
81-93-6
C18H15ClN4
322.8
SOUHUMACVWVDME-UHFFFAOYSA-N
phenosafranin,phenosafranine,phenosafranine, chloride,phenosafrinine,phenosofranine,phenosofrinine,safranin b extra,safrinin b extra,3,7-diamino-5-phenylphenazinium chloride,schultz no. 958
65732
CHEBI:33601
10-phenylphenazin-10-ium-2,8-diamine;chloride
C1=CC=C(C=C1)[N+]2=C3C=C(C=CC3=NC4=C2C=C(C=C4)N)N.[Cl-]
81-93-6
C18H15ClN4
322.8
SOUHUMACVWVDME-UHFFFAOYSA-N
phenosafranin,phenosafranine,phenosafranine, chloride,phenosafrinine,phenosofranine,phenosofrinine,safranin b extra,safrinin b extra,3,7-diamino-5-phenylphenazinium chloride,schultz no. 958
65732
CHEBI:33601
10-phenylphenazin-10-ium-2,8-diamine;chloride
C1=CC=C(C=C1)[N+]2=C3C=C(C=CC3=NC4=C2C=C(C=C4)N)N.[Cl-]

Indicator

Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Melting Point >300.0°C
Merck Index 15,7363
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 322.8
Chemical Name or Material Phenosafranin

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 201-387-