Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenosafranin, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
81-93-6
C18H15ClN4
322.796
MFCD00036335
SOUHUMACVWVDME-UHFFFAOYSA-N
phenosafranin,phenosafranine,phenosafranine, chloride,phenosafrinine,phenosofranine,phenosofrinine,safranin b extra,safrinin b extra,3,7-diamino-5-phenylphenazinium chloride,schultz no. 958
65732
CHEBI:33601
10-phenylphenazin-10-ium-2,8-diamine;chloride
C1=CC=C(C=C1)[N+]2=C3C=C(C=CC3=NC4=C2C=C(C=C4)N)N.[Cl-]
81-93-6
C18H15ClN4
322.796
MFCD00036335
SOUHUMACVWVDME-UHFFFAOYSA-N
phenosafranin,phenosafranine,phenosafranine, chloride,phenosafrinine,phenosofranine,phenosofrinine,safranin b extra,safrinin b extra,3,7-diamino-5-phenylphenazinium chloride,schultz no. 958
65732
CHEBI:33601
10-phenylphenazin-10-ium-2,8-diamine;chloride
C1=CC=C(C=C1)[N+]2=C3C=C(C=CC3=NC4=C2C=C(C=C4)N)N.[Cl-]
Indicator
Packaging Glass bottle
Infrared Spectrum Authentic
Color Green
Quantity 5g
Melting Point >300.0°C
Merck Index 15, 7363
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 322.8
Physical Form Crystalline Powder
Chemical Name or Material Phenosafranin

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.