Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenothiazine, 99%, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Plastic Bottle
Plastic drum
92-84-2
C12H9NS
199.28
MFCD00005015
WJFKNYWRSNBZNX-UHFFFAOYSA-N
phenothiazine,thiodiphenylamine,feeno,dibenzothiazine,phenosan,phenthiazine,dibenzo-1,4-thiazine,penthazine,souframine,agrazine
7108
CHEBI:37931
10H-phenothiazine
C1=CC=C2C(=C1)NC3=CC=CC=C3S2
92-84-2
C12H9NS
199.28
MFCD00005015
WJFKNYWRSNBZNX-UHFFFAOYSA-N
phenothiazine,thiodiphenylamine,feeno,dibenzothiazine,phenosan,phenthiazine,dibenzo-1,4-thiazine,penthazine,souframine,agrazine
7108
CHEBI:37931
10H-phenothiazine
C1=CC=C2C(=C1)NC3=CC=CC=C3S2
Melting Point 185.0°C
Color Yellow
Flash Point >100°C
Packaging Plastic Bottle
Infrared Spectrum Authentic
Boiling Point 371.0°C
Quantity 1kg
Assay Percent Range 99%
Beilstein 27, 63
Merck Index 15, 7364
Solubility Information Solubility in water: 2mg/l (25°C). Other solubilities: freely soluble in hot acetic acid, freely soluble in benzene and ether, slightly soluble in alcohol and mineral oils, practically insoluble in chloroform and petroleum, ether
Formula Weight 199.28
Physical Form Prills or Beads
Percent Purity 99%
Chemical Name or Material Phenothiazine
Show More Show Less

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning