Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxazine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
135-67-1
C12H9NO
183.21
MFCD00005014
TZMSYXZUNZXBOL-UHFFFAOYSA-N
phenoxazine,phenazoxine,unii-c2zwt499sg,c2zwt499sg,5,4-oxazine,phenotazine,rarechem fh 2c 0s92,phenoxazine 1g,9ci
67278
10H-phenoxazine
C1=CC=C2C(=C1)NC3=CC=CC=C3O2
135-67-1
C12H9NO
183.21
MFCD00005014
TZMSYXZUNZXBOL-UHFFFAOYSA-N
phenoxazine,phenazoxine,unii-c2zwt499sg,c2zwt499sg,5,4-oxazine,phenotazine,rarechem fh 2c 0s92,phenoxazine 1g,9ci
67278
10H-phenoxazine
C1=CC=C2C(=C1)NC3=CC=CC=C3O2
Infrared Spectrum Authentic
Melting Point 154.0°C to 159.0°C
Quantity 25g
Packaging Glass bottle
Color Gray to Gray-Green
Assay Percent Range 97%
Beilstein 27, 62
Merck Index 15, 7365
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in abs.methanol, ethanol, ether,, chloroform and benzene, sparingly soluble in petroleum ether
Formula Weight 183.21
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material Phenoxazine
Show More Show Less