Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxy resin, average M.W. 85000, Acros Organics

Click to view available options
Quantity:
250g
26402-79-9
MFCD00134004
phenoxy resin,3-4-6-propan-2-ylidene cyclohexa-2,4-dien-1-yl phenoxy propane-1,2-diol
26402-79-9
MFCD00134004
phenoxy resin,3-4-6-propan-2-ylidene cyclohexa-2,4-dien-1-yl phenoxy propane-1,2-diol
Quantity 250g
Density 1.18g/mL
Linear Formula {-C6H4C(CH3)2C6H4OCH2CH(OH)CH2O-}n
Chemical Name or Material Phenoxy resin, average M.W. 85000, average M.W. 85000