Alfa Aesar™ Phenoxyacetonitrile, 98%

Manufacturer: Alfa Aesar™ A11981-14

Quantity 25g

 View more versions of this product

Catalog No. AAA1198114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3598-14-9
    Molecular Formula C8H7NO
    Formula Weight 133.15g/mol
    MDL Number MFCD00052017
    UN Number UN3276
    Beilstein 2206192
    Chemical Name or Material Phenoxyacetonitrile

    Specifications

    Boiling Point 76° to 78°C (0.05mmHg)
    Quantity 25g
    CAS 3598-14-9
    Molecular Formula C8H7NO
    Formula Weight 133.15g/mol
    MDL Number MFCD00052017
    UN Number UN3276
    Beilstein 2206192
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Chemical Name or Material Phenoxyacetonitrile
    Assay 98%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit