Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenserine, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
101246-66-6
C20H23N3O2
337.423
PBHFNBQPZCRWQP-QUCCMNQESA-N
phenserine,unii-sue285ug3s,--eseroline phenylcarbamate,posiphen,sue285ug3s,chembl74926,--n-phenylcarbamoyleseroline,phenserine.tartaric acid,1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-pyrrolo 2,3-b indol-5-ol phenylcarbamate ester,pyrrolo 2,3-b indol-5-ol, 1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-, phenylcarbamate ester , 3as-cis
192706
[(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-phenylcarbamate
CC12CCN(C1N(C3=C2C=C(C=C3)OC(=O)NC4=CC=CC=C4)C)C
This item is not returnable. View return policy
101246-66-6
C20H23N3O2
337.423
PBHFNBQPZCRWQP-QUCCMNQESA-N
phenserine,unii-sue285ug3s,--eseroline phenylcarbamate,posiphen,sue285ug3s,chembl74926,--n-phenylcarbamoyleseroline,phenserine.tartaric acid,1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-pyrrolo 2,3-b indol-5-ol phenylcarbamate ester,pyrrolo 2,3-b indol-5-ol, 1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethyl-, phenylcarbamate ester , 3as-cis
192706
[(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-phenylcarbamate
CC12CCN(C1N(C3=C2C=C(C=C3)OC(=O)NC4=CC=CC=C4)C)C

Cholinesterase inhibitor

Physostigmine analog that inhibits acetylcholinesterase. Inhibits production of amyloid precursor protein (APP) and Aβ. Improves morris water maze performance of scopolamine-treated rats.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 337.42
Percent Purity >98%
Chemical Name or Material Phenserine

Recommended Storage : Store at +4°C