Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl 2-pyridyl ketoxime, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1826-28-4
C12H10N2O
198.225
MFCD00006310
WBSIGZZXFQOEJK-VAWYXSNFSA-N
phenylpyridylketoxime,phenyl pyridin-2-yl methanone oxime,phenyl 2-pyridyl ketone oxime,methanone, phenyl-2-pyridinyl-, oxime,2-benzoylpyridiine ketoxime,phenyl 2-pyridyl methanone oxime,2-benzoylpyridine oxime,e-phenyl-2-pyridyl ketone oxime,hydroxyimino phenyl-2-pyridylmethane,ketone, phenyl 2-pyridyl, oxime 8ci
5397197
(2E)-2-[nitroso(phenyl)methylidene]-1H-pyridine
C1=CC=C(C=C1)C(=C2C=CC=CN2)N=O
1826-28-4
C12H10N2O
198.225
MFCD00006310
WBSIGZZXFQOEJK-VAWYXSNFSA-N
phenylpyridylketoxime,phenyl pyridin-2-yl methanone oxime,phenyl 2-pyridyl ketone oxime,methanone, phenyl-2-pyridinyl-, oxime,2-benzoylpyridiine ketoxime,phenyl 2-pyridyl methanone oxime,2-benzoylpyridine oxime,e-phenyl-2-pyridyl ketone oxime,hydroxyimino phenyl-2-pyridylmethane,ketone, phenyl 2-pyridyl, oxime 8ci
5397197
(2E)-2-[nitroso(phenyl)methylidene]-1H-pyridine
C1=CC=C(C=C1)C(=C2C=CC=CN2)N=O
Color Pink
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 154.0°C to 156.0°C
Quantity 5g
Assay Percent Range 98%
Formula Weight 198.22
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Phenyl 2-pyridyl ketoxime

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.