Alfa Aesar™ Phenyl 2-pyridyl ketoxime, 99%

Manufacturer: Alfa Aesar™ H5342106

Quantity 5g

 View more versions of this product

Catalog No. AAH5342106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1826-28-4
    Molecular Formula C12H10N2O
    Formula Weight 198.22
    MDL Number MFCD00006310
    UN Number UN2811
    Beilstein 142387
    Synonym 2-Benzoylpyridine ketoxime
    Chemical Name or Material Phenyl 2-pyridyl ketoxime

    Specifications

    Formula Weight 198.22
    TSCA TSCA
    Molecular Formula C12H10N2O
    MDL Number MFCD00006310
    UN Number UN2811
    Beilstein 142387
    Health Hazard 2 GHS H Statement
    H301-H318-H315-H335
    Toxic if swallowed.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    Melting Point 154° to 156°C
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 5g
    CAS 1826-28-4
    Synonym 2-Benzoylpyridine ketoxime
    Health Hazard 1 Danger
    Chemical Name or Material Phenyl 2-pyridyl ketoxime
    Assay 99%

    GHS H Statement
    H301-H318-H315-H335
    Toxic if swallowed.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit