Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl acetate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
Packaging:
Glass bottle
122-79-2
C8H8O2
136.15
MFCD00008699
IPBVNPXQWQGGJP-UHFFFAOYSA-N
acetic acid phenyl ester,phenol acetate,acetylphenol,acetic acid, phenyl ester,acetyl phenol,acetoxybenzene,acetates,acetic acid,phenyl ester,fenylester kyseliny octove,fenylester kyseliny octove czech
31229
CHEBI:8082
phenyl acetate
CC(=O)OC1=CC=CC=C1
122-79-2
C8H8O2
136.15
MFCD00008699
IPBVNPXQWQGGJP-UHFFFAOYSA-N
acetic acid phenyl ester,phenol acetate,acetylphenol,acetic acid, phenyl ester,acetyl phenol,acetoxybenzene,acetates,acetic acid,phenyl ester,fenylester kyseliny octove,fenylester kyseliny octove czech
31229
CHEBI:8082
phenyl acetate
CC(=O)OC1=CC=CC=C1
Refractive Index 1.5010 to 1.5030
Color Colorless to Brown
Quantity 5g
Specific Gravity 1.07
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.0700g/mL
Boiling Point 196.0°C
Flash Point 76°C
Assay Percent Range 96% min. (GC)
Linear Formula CH3CO2C6H5
Beilstein 06,152
Fieser 01,829
Merck Index 15,7379
Solubility Information Solubility in water: ca 4g/L (25°C). Other solubilities: miscible with alcohol,chloroform,ether,soluble in glacial acetic acid
Formula Weight 136.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Phenyl acetate, 97%
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 204-575-0

RTECSNumber : AJ2800000

TSCA : TSCA