Alfa Aesar™ Phenyl benzoate, 99%

Manufacturer: Alfa Aesar™ B2298636

Quantity 500g

 View more versions of this product

Catalog No. AAB2298636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 93-99-2
    Molecular Formula C13H10O2
    Formula Weight 198.22
    MDL Number MFCD00003072
    Merck Index 14,7275
    Synonym Benzoic acid phenyl ester
    Chemical Name or Material Phenyl benzoate

    Specifications

    Formula Weight 198.22
    MDL Number MFCD00003072
    CAS 93-99-2
    Synonym Benzoic acid phenyl ester
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DH6299500
    Boiling Point 298° to 299°C
    Melting Point 68° to 70°C
    TSCA TSCA
    Molecular Formula C13H10O2
    Merck Index 14,7275
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Quantity 500g
    Chemical Name or Material Phenyl benzoate
    Assay 99%

    Description

    Applications
    Phenyl benzoate was used in the synthesis of soluble polyimides using dianhydride/diamine derivatives. It undergoes Fries rearrangement catalyzed by heteropoly acids to yield the acylated phenols and esters. Phenyl benzoate is considered an excellent starting material for the production of optical components, particularly high quality lenses for still and motion picture cameras.

    Solubility
    Soluble in ethanol, ethyl ether and chloroform. Insoluble in water.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit