Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl bromoacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
620-72-4
C8H7BrO2
215.046
MFCD00192391
UEWYUCGVQMZMGY-UHFFFAOYSA-N
phenyl bromoacetate,acetic acid, bromo-, phenyl ester,bromoacetic acid phenyl ester,bromoacetic acid, phenyl ester,phenylbromessigester,phenyl bromoacetate #,acmc-209mzb,phenyl 2-bromanylethanoate,ksc352o8b
564919
phenyl 2-bromoacetate
C1=CC=C(C=C1)OC(=O)CBr
620-72-4
C8H7BrO2
215.046
MFCD00192391
UEWYUCGVQMZMGY-UHFFFAOYSA-N
phenyl bromoacetate,acetic acid, bromo-, phenyl ester,bromoacetic acid phenyl ester,bromoacetic acid, phenyl ester,phenylbromessigester,phenyl bromoacetate #,acmc-209mzb,phenyl 2-bromanylethanoate,ksc352o8b
564919
phenyl 2-bromoacetate
C1=CC=C(C=C1)OC(=O)CBr
Quantity 50g
Density 1.508g/mL
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 26°C to 33°C
Assay Percent Range 98%
Formula Weight 215.05
Physical Form Low Melting Solid or Liquid
Percent Purity ≥97.5%
Chemical Name or Material Phenyl bromoacetate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.