Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl chlorothionocarbonate, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1005-56-7
C7H5ClOS
172.63
KOSYAAIZOGNATQ-UHFFFAOYSA-N
o-phenyl carbonochloridothioate,phenyl chlorothionoformate,o-phenyl chlorothioformate,phenyl chlorothioformate,phenyl thioxochloroformate,phenyl chlorothionocarbonate,phenoxythiocarbonyl chloride,chlorothioformic acid phenyl ester,o-phenyl chlorothionoformate,o-phenyl chlorothiocarbonate
70498
O-phenyl chloromethanethioate
C1=CC=C(C=C1)OC(=S)Cl
1005-56-7
C7H5ClOS
172.63
KOSYAAIZOGNATQ-UHFFFAOYSA-N
o-phenyl carbonochloridothioate,phenyl chlorothionoformate,o-phenyl chlorothioformate,phenyl chlorothioformate,phenyl thioxochloroformate,phenyl chlorothionocarbonate,phenoxythiocarbonyl chloride,chlorothioformic acid phenyl ester,o-phenyl chlorothionoformate,o-phenyl chlorothiocarbonate
70498
O-phenyl chloromethanethioate
C1=CC=C(C=C1)OC(=S)Cl
Specific Gravity 1.248
Packaging Glass bottle
Density 1.2480g/mL
Flash Point 81°C
Color Yellow
Boiling Point 81.0°C to 83.0°C (6.0mmHg)
Quantity 25g
Refractive Index 1.4795 to 1.4815
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula ClC(S)OC6H5
Beilstein 06,III,609
Fieser 10,306
Solubility Information Solubility in water: decomposes
Formula Weight 172.63
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Phenyl chlorothionocarbonate
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 213-736-4