Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl ether, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
101-84-8
C12H10O
170.21
MFCD00003034
USIUVYZYUHIAEV-UHFFFAOYSA-N
diphenyl ether,diphenyl oxide,phenyl ether,oxydibenzene,phenyl oxide,oxybisbenzene,biphenyl oxide,1,1'-oxydibenzene,benzene, 1,1'-oxybis,oxydiphenyl
7583
CHEBI:39258
phenoxybenzene
C1=CC=C(C=C1)OC2=CC=CC=C2
101-84-8
C12H10O
170.21
MFCD00003034
USIUVYZYUHIAEV-UHFFFAOYSA-N
diphenyl ether,diphenyl oxide,phenyl ether,oxydibenzene,phenyl oxide,oxybisbenzene,biphenyl oxide,1,1'-oxydibenzene,benzene, 1,1'-oxybis,oxydiphenyl
7583
CHEBI:39258
phenoxybenzene
C1=CC=C(C=C1)OC2=CC=CC=C2
Specific Gravity 1.073
Viscosity 3.4909 mPa.s (28°C)
Melting Point 26.0°C to 30.0°C
Quantity 10kg
Flash Point 115°C
Refractive Index 1.5795 to 1.5815
Packaging Plastic drum
Density 1.0730g/mL
Boiling Point 259.0°C
Assay Percent Range 99%
Linear Formula (C6H5)2O
Beilstein 06,146
Merck Index 15,3357
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether and,glacial acetic acid
Formula Weight 170.21
Percent Purity 99%
Chemical Name or Material Phenyl ether
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning