Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Ether-biphenyl Eutectic, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
8004-13-5
C24H20O
324.41
MFCD00148859
MHCVCKDNQYMGEX-UHFFFAOYSA-N
diphyl, dowtherm, dowtherm a, dinil, dinyl, therminol vp, phenyl ether-biphenyl mixture, phenyl ether-diphenyl mixture, biphenyl-diphenyl ether mixture, hsdb 137
24670
1,1'-biphenyl;phenoxybenzene
C1=CC=C(C=C1)C2=CC=CC=C2.C1=CC=C(C=C1)OC2=CC=CC=C2
8004-13-5
C24H20O
324.41
MFCD00148859
MHCVCKDNQYMGEX-UHFFFAOYSA-N
diphyl, dowtherm, dowtherm a, dinil, dinyl, therminol vp, phenyl ether-biphenyl mixture, phenyl ether-diphenyl mixture, biphenyl-diphenyl ether mixture, hsdb 137
24670
1,1'-biphenyl;phenoxybenzene
C1=CC=C(C=C1)C2=CC=CC=C2.C1=CC=C(C=C1)OC2=CC=CC=C2
Packaging Glass bottle
Refractive Index 1.5885 to 1.5905
Specific Gravity 1.056
Flash Point 123°C
Melting Point 13°C
Density 1.0560g/mL
Quantity 100g
Formula Weight 324.41
Chemical Name or Material Phenyl ether-biphenyl eutectic

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

RTECSNumber : DV1500000