Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Isocyanate 99%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
Packaging:
Glass bottle
103-71-9
C7H5NO
119.12
MFCD00001994
DGTNSSLYPYDJGL-UHFFFAOYSA-N
phenyl isocyanate,phenylcarbimide,carbanil,phenyl carbonimide,benzene, isocyanato,phenylisocyanate,isocyanic acid, phenyl ester,mondur p,phnco,fenylisokyanat
7672
CHEBI:53806
isocyanatobenzene
C1=CC=C(C=C1)N=C=O
103-71-9
C7H5NO
119.12
MFCD00001994
DGTNSSLYPYDJGL-UHFFFAOYSA-N
phenyl isocyanate,phenylcarbimide,carbanil,phenyl carbonimide,benzene, isocyanato,phenylisocyanate,isocyanic acid, phenyl ester,mondur p,phnco,fenylisokyanat
7672
CHEBI:53806
isocyanatobenzene
C1=CC=C(C=C1)N=C=O
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5350 to 1.5370
Color Colorless to Yellow
Quantity 1kg
Density 1.0950g/mL
Melting Point -31.3°C
Boiling Point 162.0°C to 163.0°C
Flash Point 51°C
Specific Gravity 1.095
Assay Percent Range 99%
Linear Formula C6H5NCO
Beilstein 12, 437
Fieser 01,842; 02,322; 04,378; 07,284; 08,396; 10,309; 12,386
Merck Index 15, 7407
Solubility Information Solubility in water: decomposes. Other solubilities: decomposes in alcohol, soluble in ether
Formula Weight 119.12
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Phenyl isocyanate, 99%
Show More Show Less

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Fatal if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-137-6

RTECSNumber : DA3675000

TSCA : TSCA