Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl N-phenylphosphoramidochloridate, ≥99%, Acros Organics

Click to view available options
Quantity:
5g
51766-21-3
C12H11ClNO2P
267.649
MFCD00003029
ZUQYQPGYEFBITH-UHFFFAOYSA-N
phenyl phenylphosphoramidochloridate,phenyl n-phenylphosphoramidochloridate,3p-cl,phenoxy phenylamino phosphinoyl chloride,acmc-20ajx0,zuqyqpgyefbith-uhfffaoysa,phenyl-n-phenylchlorophosphoramidate,n-chloro phenoxy phosphoryl aniline,phenyl n-phenyl-chlorophosphoramidate,n-chloro-phenoxy phosphoryl aniline
103959
N-[chloro(phenoxy)phosphoryl]aniline
C1=CC=C(C=C1)NP(=O)(OC2=CC=CC=C2)Cl
51766-21-3
C12H11ClNO2P
267.649
MFCD00003029
ZUQYQPGYEFBITH-UHFFFAOYSA-N
phenyl phenylphosphoramidochloridate,phenyl n-phenylphosphoramidochloridate,3p-cl,phenoxy phenylamino phosphinoyl chloride,acmc-20ajx0,zuqyqpgyefbith-uhfffaoysa,phenyl-n-phenylchlorophosphoramidate,n-chloro phenoxy phosphoryl aniline,phenyl n-phenyl-chlorophosphoramidate,n-chloro-phenoxy phosphoryl aniline
103959
N-[chloro(phenoxy)phosphoryl]aniline
C1=CC=C(C=C1)NP(=O)(OC2=CC=CC=C2)Cl
Infrared Spectrum Authentic
Quantity 5g
Color White
Melting Point 132.5°C to 134.5°C
Assay Percent Range 99+%
Linear Formula C6H5NHP(O)(Cl)OC6H5
Beilstein 12, 588
Fieser 07,286; 10,310; 11,416
Formula Weight 267.64
Physical Form Powder
Percent Purity ≥99.0%
Chemical Name or Material Phenyl N-phenylphosphoramidochloridate, 99+ %
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE