Alfa Aesar™ Phenyl nicotinate, 99%

Manufacturer: Alfa Aesar™ B2221914

Quantity 25g

 View more versions of this product

Catalog No. AAB2221914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3468-53-9
    Molecular Formula C12H9NO2
    Formula Weight 199.21
    MDL Number MFCD00023581
    Beilstein 140968
    Synonym Nicotinic acid phenyl ester; Phenyl pyridine-3-carboxylate
    Chemical Name or Material Phenyl nicotinate

    Specifications

    Formula Weight 199.21
    TSCA TSCA
    Molecular Formula C12H9NO2
    MDL Number MFCD00023581
    Synonym Nicotinic acid phenyl ester; Phenyl pyridine-3-carboxylate
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H317-H335-H315
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 186° to 188°C (20mmHg)
    Melting Point 70° to 74°C
    Quantity 25g
    CAS 3468-53-9
    Beilstein 140968
    Chemical Name or Material Phenyl nicotinate
    Assay 99%

    Description

    Applications
    In nicotinic acid analogs assayed against tri-o-cresyl phosphate-induced ataxia in hens Phenyl nicotinate provided partial relief of the syndrome.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H318-H317-H335-H315
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit