Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phenyl nicotinate, 99%

Click to view available options
Quantity:
5g
25g
100g
3468-53-9
C12H9NO2
199.209
MFCD00023581
QEJPOSAIULNDLU-UHFFFAOYSA-N
phenyl nicotinate,3-pyridinecarboxylic acid, phenyl ester,phenylnicotinate,phenyl 3-pyridinecarboxylate,nicotinic acid, phenyl ester,unii-vrd2l6rm16,vrd2l6rm16,acmc-209i9k,nicotinic acid phenyl ester,qejposaiulndlu-uhfffaoysa
77018
phenyl pyridine-3-carboxylate
C1=CC=C(C=C1)OC(=O)C2=CN=CC=C2
3468-53-9
C12H9NO2
199.209
MFCD00023581
QEJPOSAIULNDLU-UHFFFAOYSA-N
phenyl nicotinate,3-pyridinecarboxylic acid, phenyl ester,phenylnicotinate,phenyl 3-pyridinecarboxylate,nicotinic acid, phenyl ester,unii-vrd2l6rm16,vrd2l6rm16,acmc-209i9k,nicotinic acid phenyl ester,qejposaiulndlu-uhfffaoysa
77018
phenyl pyridine-3-carboxylate
C1=CC=C(C=C1)OC(=O)C2=CN=CC=C2
Applications
In nicotinic acid analogs assayed against tri-o-cresyl phosphate-induced ataxia in hens Phenyl nicotinate provided partial relief of the syndrome.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Quantity 5g
Melting Point 70°C to 74°C
Boiling Point 186°C to 188°C (20mmHg)
Beilstein 140968
Formula Weight 199.21
Percent Purity 99%
Chemical Name or Material Phenyl nicotinate

GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA