Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Phenyl nicotinate, 99%
Click to view available options
Quantity:
5g
25g
100g
Description
Applications
In nicotinic acid analogs assayed against tri-o-cresyl phosphate-induced ataxia in hens Phenyl nicotinate provided partial relief of the syndrome.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
In nicotinic acid analogs assayed against tri-o-cresyl phosphate-induced ataxia in hens Phenyl nicotinate provided partial relief of the syndrome.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Quantity | 100g |
Boiling Point | 186°C to 188°C (20mmHg) |
Melting Point | 70°C to 74°C |
Beilstein | 140968 |
Formula Weight | 199.21 |
Percent Purity | 99% |
Chemical Name or Material | Phenyl nicotinate |
Safety and Handling
GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c
H315-H317-H318-H335
EINECSNumber : 222-428-9
TSCA : Yes
Recommended Storage : Ambient temperatures