Alfa Aesar™ Phenyl phosphate disodium salt hydrate, 98%

Used in phosphatase determinations

Manufacturer: Alfa Aesar™ 4471122

Quantity 100g

 View more versions of this product

Catalog No. AA4471122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3279-54-7
    Molecular Formula C6H5Na2O4P·xH2O
    Linear Formula C6H5OPO(ONa)2·xH2O
    Formula Weight 218.06 (Anhydrous)
    MDL Number MFCD00002133
    Beilstein 4167154
    Merck Index 14,3357
    Chemical Name or Material Phenyl phosphate disodium salt hydrate

    Specifications

    Formula Weight 218.06 (Anhydrous)
    MDL Number MFCD00002133
    CAS 3279-54-7
    Physical Form Crystalline
    TSCA TSCA
    Molecular Formula C6H5Na2O4P·xH2O
    Linear Formula C6H5OPO(ONa)2·xH2O
    Sensitivity Hygroscopic
    Quantity 100g
    Beilstein 4167154
    Merck Index 14,3357
    Color Pale yellow
    Chemical Name or Material Phenyl phosphate disodium salt hydrate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit