Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phenyl phosphate disodium salt hydrate, 98%

Used in phosphatase determinations

Manufacturer:  Alfa Aesar™ 4471114

Quantity 25g

 View more versions of this product

Catalog No. AA4471114


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

3279-54-7
C6H5OPO(ONa)2·xH2O
MFCD00002133
14,3357
C6H5Na2O4P·xH2O
218.06 (Anhydrous)
4167154
Phenyl phosphate disodium salt hydrate

Specifications

Hygroscopic
3279-54-7
C6H5Na2O4P·xH2O
218.06 (Anhydrous)
4167154
Pale yellow
98%
25g
Crystalline
C6H5OPO(ONa)2·xH2O
MFCD00002133
14,3357
Phenyl phosphate disodium salt hydrate

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit