Alfa Aesar™ Phenyl phosphorodichloridate, 97%

Manufacturer: Alfa Aesar™ A1047936

Quantity 500g

 View more versions of this product

Catalog No. AAA1047936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 770-12-7
    Molecular Formula C6H5Cl2O2P
    Formula Weight 210.98
    MDL Number MFCD00002067
    UN Number UN3265
    Beilstein 511863
    Synonym Phenyl dichlorophosphate
    Chemical Name or Material Phenyl phosphorodichloridate

    Specifications

    Formula Weight 210.98
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H5Cl2O2P
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Melting Point −1°C
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    TSCA TSCA
    MDL Number MFCD00002067
    CAS 770-12-7
    UN Number UN3265
    Beilstein 511863
    Boiling Point 241° to 243°C
    RTECS Number TD4393000
    Synonym Phenyl dichlorophosphate
    Health Hazard 1 Danger
    Flash Point 112°C (233°F)
    Quantity 500g
    Chemical Name or Material Phenyl phosphorodichloridate
    Assay 97%

    Description

    Applications
    Reagent for preparation of phosphate diesters.Phenyl phosphorodichloridate is used in the preparation of symmetrical phosphate diesters. It is used as a phosphorylating agent for alcohols and amines. It is also involved in the preparation of phenylphosphoro di-(1-imidazolidate) and 2-phenyl-bis-triazoloylphosphate, which is used in peptide synthesis. It plays a vital role in the Pfitzner-Moffatt oxidation which involves oxidation of alcohols to the corresponding aldehydes and ketones. Furthermore, it is utilized in Beckmann rearrangement for the preparation of N-phenylacetamide from acetophenone oxime.

    Solubility
    Immiscible with water.

    Notes
    Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, alcohols and strong oxidizing agents.

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit