Alfa Aesar™ Phenyl propargyl ether, 97%

Manufacturer: Alfa Aesar™ 4392322

Quantity 100g

 View more versions of this product

Catalog No. AA4392322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13610-02-1
    Molecular Formula C9H8O
    Linear Formula C6H5OCH2C≡CH
    Formula Weight 132.16
    MDL Number MFCD00048107
    Synonym 3-Phenoxy-1-propyne
    Chemical Name or Material Phenyl propargyl ether

    Specifications

    Formula Weight 132.16
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula C6H5OCH2C≡CH
    Density 1.030g/mL
    Synonym 3-Phenoxy-1-propyne
    Refractive Index 1.5330
    Boiling Point 89° to 90°C (14mmHg)
    Physical Form Liquid
    MDL Number MFCD00048107
    CAS 13610-02-1
    Health Hazard 1 Danger
    Quantity 100g
    Molecular Formula C9H8O
    Health Hazard 2 GHS H Statement
    H318-H335-H227-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Combustible liquid.
    Causes skin irritation.
    Odor Pungent
    Color Colorless to yellow
    Chemical Name or Material Phenyl propargyl ether
    Assay 97%

    GHS H Statement
    H318-H335-H227-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Combustible liquid.
    Causes skin irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit