Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Sulfone 97%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
Packaging:
Glass Bottle
Plastic bottle
127-63-9
C12H10O2S
218.28
MFCD00007548
KZTYYGOKRVBIMI-UHFFFAOYSA-N
diphenyl sulfone,phenyl sulfone,sulfonyldibenzene,diphenylsulfone,sulfobenzide,diphenyl sulphone,benzene, 1,1'-sulfonylbis,sulfone, diphenyl,1,1'-sulfonyldibenzene,phenyl sulphone
31386
CHEBI:78360
benzenesulfonylbenzene
C1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2
127-63-9
C12H10O2S
218.28
MFCD00007548
KZTYYGOKRVBIMI-UHFFFAOYSA-N
diphenyl sulfone,phenyl sulfone,sulfonyldibenzene,diphenylsulfone,sulfobenzide,diphenyl sulphone,benzene, 1,1'-sulfonylbis,sulfone, diphenyl,1,1'-sulfonyldibenzene,phenyl sulphone
31386
CHEBI:78360
benzenesulfonylbenzene
C1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 125.0°C to 129.0°C
Color White
Quantity 1kg
Boiling Point 379.0°C
Assay Percent Range 97%
Linear Formula (C6H5)2SO2
Beilstein 06, 300
Merck Index 15, 3372
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in boiling water,soluble in hot alcohol and benzene
Formula Weight 218.28
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Phenyl sulfone, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-853-1

RTECSNumber : SX2400000

TSCA : TSCA