Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl tosylate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
640-60-8
C13H12O3S
248.296
MFCD00025996
KZQFPRKQBWRRHQ-UHFFFAOYSA-N
phenyl para-toluenesulfonate,p-toluenesulfonic acid phenyl ester,phenyl tosylate,phenyl p-toluenesulfonate,phenylp-toluenesulfonate,acmc-209njz,phenyl 4-methylbenzene-1-sulfonate,toluene-4-sulfonic acid phenyl ester,benzenesulfonic acid,4-methyl-, phenyl ester
223146
phenyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OC2=CC=CC=C2
640-60-8
C13H12O3S
248.296
MFCD00025996
KZQFPRKQBWRRHQ-UHFFFAOYSA-N
phenyl para-toluenesulfonate,p-toluenesulfonic acid phenyl ester,phenyl tosylate,phenyl p-toluenesulfonate,phenylp-toluenesulfonate,acmc-209njz,phenyl 4-methylbenzene-1-sulfonate,toluene-4-sulfonic acid phenyl ester,benzenesulfonic acid,4-methyl-, phenyl ester
223146
phenyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OC2=CC=CC=C2
Color White to Beige
Quantity 25g
Packaging Glass bottle
Melting Point 94°C to 98°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 248.3
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Phenyl tosylate, 98%

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.