Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetaldehyde Dimethyl Acetal 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
101-48-4
C10H14O2
166.22
MFCD00008487
WNJSKZBEWNVKGU-UHFFFAOYSA-N
2,2-dimethoxyethyl benzene,phenylacetaldehyde dimethyl acetal,viridine,hyscylene p,1,1-dimethoxy-2-phenylethane,benzene, 2,2-dimethoxyethyl,phenacetaldehyde dimethyl acetal,padma,2,2-dimethoxy-1-phenylethane,phenylacetic aldehyde dimethyl acetal
60995
2,2-dimethoxyethylbenzene
COC(CC1=CC=CC=C1)OC
101-48-4
C10H14O2
166.22
MFCD00008487
WNJSKZBEWNVKGU-UHFFFAOYSA-N
2,2-dimethoxyethyl benzene,phenylacetaldehyde dimethyl acetal,viridine,hyscylene p,1,1-dimethoxy-2-phenylethane,benzene, 2,2-dimethoxyethyl,phenacetaldehyde dimethyl acetal,padma,2,2-dimethoxy-1-phenylethane,phenylacetic aldehyde dimethyl acetal
60995
2,2-dimethoxyethylbenzene
COC(CC1=CC=CC=C1)OC
Density 1.0040g/mL
Specific Gravity 1.004
Refractive Index 1.4930 to 1.495
Boiling Point 219°C to 221°C (754.0mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Quantity 5g
Acidity 1mg KOH/g max.
Color Colorless to Yellow
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula C6H5CH2CH(OCH3)2
Beilstein 07, 293
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 2 parts of 70% alcohol
Formula Weight 166.22
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Phenylacetaldehyde dimethyl acetal, 98%
Show More Show Less

EINECSNumber : 202-945-6

RTECSNumber : AB3040000

TSCA : TSCA