Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetic acid hydrazide, 98%, Acros Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
937-39-3
C8H10N2O
150.18
MFCD00007612
FPTCVTJCJMVIDV-UHFFFAOYSA-N
phenylacetic acid hydrazide,benzeneacetic acid, hydrazide,phenylacetic hydrazide,phenylacetylhydrazine,phenylacetyl hydrazide,phenacetic acid hydrazide,2-phenylacetyl hydrazine,phenylacetohydrazide,phenylacethydrazide,phenyl-acetic acid hydrazide
70301
2-phenylacetohydrazide
C1=CC=C(C=C1)CC(=O)NN
937-39-3
C8H10N2O
150.18
MFCD00007612
FPTCVTJCJMVIDV-UHFFFAOYSA-N
phenylacetic acid hydrazide,benzeneacetic acid, hydrazide,phenylacetic hydrazide,phenylacetylhydrazine,phenylacetyl hydrazide,phenacetic acid hydrazide,2-phenylacetyl hydrazine,phenylacetohydrazide,phenylacethydrazide,phenyl-acetic acid hydrazide
70301
2-phenylacetohydrazide
C1=CC=C(C=C1)CC(=O)NN
Melting Point 115.0°C to 117.0°C
Quantity 100g
Color White to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula C6H5CH2CONHNH2
Beilstein 09, 446
Formula Weight 150.18
Physical Form Needles
Percent Purity 98%
Chemical Name or Material Phenylacetic acid hydrazide
Show More Show Less