Alfa Aesar™ Phenylacetyl chloride, 98%

Manufacturer: Alfa Aesar™ B249870B

Quantity 1000g

 View more versions of this product

Catalog No. AAB249870B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 103-80-0
    Molecular Formula C8H7ClO
    Formula Weight 154.6
    MDL Number MFCD00000729
    UN Number UN2577
    Beilstein 742254
    Chemical Name or Material Phenylacetyl chloride

    Specifications

    Formula Weight 154.6
    CAS 103-80-0
    UN Number UN2577
    Beilstein 742254
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHENYLACETYL CHLORIDE
    Boiling Point 225°C
    TSCA TSCA
    Molecular Formula C8H7ClO
    MDL Number MFCD00000729
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flash Point 102°C (215°F)
    Quantity 1000g
    Chemical Name or Material Phenylacetyl chloride
    Assay 98%

    Description

    Applications
    Phenylacetyl chloride is used in the preparation of 1,2-diphenyl-ethanone by reaction with iodobenzene using tetrabutylammonium tetrafluoroborate as a phase transfer catalyst. It is widely used in the chemical industry for manufacturing dyes and pharmaceutical products. It plays an important role in the preparation of penicillin. Further, it is used to prepare esters for flavorings.

    Solubility
    Miscible with alcohol and ether.

    Notes
    Moisture sensitive. Store in a cool place.Incompatible with amines, most common metals, strong bases and strong oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit