Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetyl chloride 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
103-80-0
C8H7ClO
154.593
MFCD00000729
VMZCDNSFRSVYKQ-UHFFFAOYSA-N
phenylacetyl chloride,benzeneacetyl chloride,acetyl chloride, phenyl,phenylacetic acid chloride,phenacetyl chloride,phenylacetylchloride,unii-t30899drnd,phenyl acetyl chloride,a-toluoyl chloride,phenylacetyl
7679
2-phenylacetyl chloride
C1=CC=C(C=C1)CC(=O)Cl
103-80-0
C8H7ClO
154.593
MFCD00000729
VMZCDNSFRSVYKQ-UHFFFAOYSA-N
phenylacetyl chloride,benzeneacetyl chloride,acetyl chloride, phenyl,phenylacetic acid chloride,phenacetyl chloride,phenylacetylchloride,unii-t30899drnd,phenyl acetyl chloride,a-toluoyl chloride,phenylacetyl
7679
2-phenylacetyl chloride
C1=CC=C(C=C1)CC(=O)Cl
Refractive Index 1.532 to 1.533
Density 1.16
Flash Point 102°C
Quantity 5g
Boiling Point 94°C to 95°C (12mmHg)
Solubility Information Solubility in water: Reacts
Formula Weight 154.6
Chemical Name or Material Phenylacetyl chloride 98%, 98%