Learn More
Phenylacetylene, 98+%, Thermo Scientific Chemicals
Description
Phenylacetylene is involved in the preparation of styrene by reduction using Lindlar catalyst. It is used to study the mechanism of the palladium-catalyzed phenyl acetylene oxidative carbonylation reaction. It is also used in the polymerization process to prepare polyphenylacetylene namely 1,2,4-triphenylbenzene and 1,3,5-triphenylbenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phenylacetylene is involved in the preparation of styrene by reduction using Lindlar catalyst. It is used to study the mechanism of the palladium-catalyzed phenyl acetylene oxidative carbonylation reaction. It is also used in the polymerization process to prepare polyphenylacetylene namely 1,2,4-triphenylbenzene and 1,3,5-triphenylbenzene.
Solubility
Immiscible with water.
Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, halogens, alkali metals and oxidizing agents.
Specifications
Melting Point | -45°C |
Density | 0.93 |
Boiling Point | 142°C to 144°C |
Flash Point | 27°C (81°F) |
Refractive Index | 1.548 |
Quantity | 25 g |
UN Number | UN3295 |
Beilstein | 605461 |
Sensitivity | Light sensitive |
Merck Index | 14,3861 |
Show More |
Safety and Handling
Hazard Category | H226-H304-H315-H319 |
Hazard Statement | GHS H Statement H304-H226-H319-H335 May be fatal if swallowed and enters airways. Flammable liquid and vapor. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
EINECSNumber | 208-645-1 |
RTECSNumber | DA0780000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.