Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetylurea 98%, Acros Organics

Click to view available options
Quantity:
100g
63-98-9
C9H10N2O2
178.191
MFCD00007948
XPFRXWCVYUEORT-UHFFFAOYSA-N
phenacemide,phenylacetylurea,phenacetylurea,phenuron,phenurone,cetylureum,fenacemid,neophedan,phenacalum,phenacetur
4753
CHEBI:8049
N-carbamoyl-2-phenylacetamide
C1=CC=C(C=C1)CC(=O)NC(=O)N
63-98-9
C9H10N2O2
178.191
MFCD00007948
XPFRXWCVYUEORT-UHFFFAOYSA-N
phenacemide,phenylacetylurea,phenacetylurea,phenuron,phenurone,cetylureum,fenacemid,neophedan,phenacalum,phenacetur
4753
CHEBI:8049
N-carbamoyl-2-phenylacetamide
C1=CC=C(C=C1)CC(=O)NC(=O)N
Quantity 100g
Assay Percent Range 98%
Linear Formula C6H5CH2CONHCONH2
Merck Index 15, 7318
Formula Weight 178.19
Chemical Name or Material Phenylacetylurea, 98%

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.