Alfa Aesar™ Phenylarsonic acid, 97%

Manufacturer: Alfa Aesar™ A1062506

Quantity 5g

 View more versions of this product

Catalog No. AAA1062506

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 98-05-5
    Molecular Formula C6H7AsO3
    Formula Weight 202.04
    MDL Number MFCD00002097
    UN Number UN3465
    Merck Index 14,1067
    Synonym Benzenearsonic acid
    Chemical Name or Material Phenylarsonic acid

    Specifications

    Formula Weight 202.04
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00002097
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Quantity 5g
    TSCA TSCA
    Molecular Formula C6H7AsO3
    CAS 98-05-5
    Merck Index 14,1067
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
    RTECS Number CY3150000
    UN Number UN3465
    Synonym Benzenearsonic acid
    Melting Point 158° to 162°C (dec.)
    Chemical Name or Material Phenylarsonic acid
    Assay 97%

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit