Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylbutazone, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
50-33-9
C19H20N2O2
308.38
MFCD00005500
VYMDGNCVAMGZFE-UHFFFAOYSA-N
phenylbutazone,fenilbutazona,butazolidin,diphenylbutazone,phenylbutazonum,butapirazol,butapyrazole,butazolidine,butacote,butadion
4781
CHEBI:48574
4-butyl-1,2-diphenylpyrazolidine-3,5-dione
CCCCC1C(=O)N(N(C1=O)C2=CC=CC=C2)C3=CC=CC=C3
50-33-9
C19H20N2O2
308.38
MFCD00005500
VYMDGNCVAMGZFE-UHFFFAOYSA-N
phenylbutazone,fenilbutazona,butazolidin,diphenylbutazone,phenylbutazonum,butapirazol,butapyrazole,butazolidine,butacote,butadion
4781
CHEBI:48574
4-butyl-1,2-diphenylpyrazolidine-3,5-dione
CCCCC1C(=O)N(N(C1=O)C2=CC=CC=C2)C3=CC=CC=C3
Color White
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 104°C to 107°C
Assay Percent Range 99+%
Merck Index 15, 7390
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in solutions of alkali hydroxides,freely soluble in acetone,ether and ethyl acetate
Formula Weight 308.38
Physical Form Powder
Percent Purity 99+%
Chemical Name or Material Phenylbutazone
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-029-