Alfa Aesar™ Phenylhydrazine, 97%

Manufacturer: Alfa Aesar™ A112460B

Quantity 1000g

 View more versions of this product

Catalog No. AAA112460B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-63-0
    Molecular Formula C6H8N2
    Formula Weight 108.14
    MDL Number MFCD00007573
    UN Number UN2572
    Beilstein 606080
    Merck Index 14,7293
    Chemical Name or Material Phenylhydrazine

    Specifications

    Formula Weight 108.14
    Assay Percent Range 97%
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H350-H372-H341-H227-H315-H319-H317 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2572
    MDL Number MFCD00007573
    Flash Point 88°C (190°F)
    Boiling Point 238° to 241°C
    Quantity 1000g
    TSCA TSCA
    Molecular Formula C6H8N2
    Beilstein 606080
    Merck Index 14,7293
    Solubility Information Soluble in dilute acids
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENYLHYDRAZINE
    RTECS Number MV8925000
    Melting Point 18° to 20°C
    CAS 100-63-0
    Chemical Name or Material Phenylhydrazine
    Assay 97%

    Description

    Applications
    Phenyl hydrazine is involved in Fischer indole synthesis to prepare indoles, which find application as intermediates in the pharmaceuticals particularly for the tryptamine drug. In analytical chemistry, it is used to differentiate and separate sugars by forming phenyhydrazones. It is used as an N-protecting reagent and for cleavage of phthaloyl group.

    Solubility
    Soluble in dilute acids.

    Notes
    Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H311-H331-H350-H372-H341-H227-H315-H319-H317 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.

    GHS P Statement
    P210-P301+P310-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit