Alfa Aesar™ Phenyllithium, typically 1.9M in di-n-butyl ether

Manufacturer: Alfa Aesar™ 45770AE

Quantity 100mL

 View more versions of this product

Catalog No. AA45770AE

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 591-51-5
    Molecular Formula C6H5Li
    Formula Weight 84.05
    MDL Number MFCD00014254
    UN Number UN2924
    Beilstein 506502
    Chemical Name or Material Phenyllithium

    Specifications

    Formula Weight 84.05
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H5Li
    CAS 591-51-5
    Health Hazard 1 Danger
    UN Number UN2924
    Beilstein 506502
    Density 0.81g/mL
    Boiling Point 140° to 143°C
    Color Dark brown to black
    Flash Point 25°C (77°F)
    Sensitivity Air & Moisture Sensitive
    Concentration Typically 1.9M in di-n-butyl ether
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00014254
    Quantity 100mL
    Health Hazard 2 GHS H Statement
    H340-H350-H314-H318-H226-H335
    May cause genetic defects.
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material Phenyllithium

    GHS H Statement
    H340-H350-H314-H318-H226-H335
    May cause genetic defects.
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    May cause respiratory irritation.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit