Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phenyllithium, typically 1.9M in di-n-butyl ether

Manufacturer:  Alfa Aesar™ 457706C

Quantity 5 x 100mL

 View more versions of this product

Catalog No. AA457706C

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

591-51-5
84.05
UN2924
Phenyllithium
C6H5Li
MFCD00014254
506502

Specifications

Typically 1.9M in di-n-butyl ether
5 x 100mL
140° to 143°C
25°C (77°F)
Liquid
84.05
UN2924
Phenyllithium
591-51-5
0.81g/mL
Dark brown to black
Air & Moisture Sensitive
C6H5Li
MFCD00014254
506502

Safety and Handling

SDS & Safety Documents

GHS H Statement
H340-H350-H314-H318-H226-H335
May cause genetic defects.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit