Alfa Aesar™ Phenylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 42859-JT

Quantity 0.5 mole

 View more versions of this product

Catalog No. AA42859JT

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 100-58-3
    Formula Weight 181.31
    MDL Number MFCD00000038
    UN Number UN3399
    Chemical Name or Material Phenylmagnesium bromide

    Specifications

    Formula Weight 181.31
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off i
    Storage Note 1
    • Packaged under Argon
    Packaging ChemSeal™ bottles
    Physical Form Liquid
    MDL Number MFCD00000038
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    Density 1.14g/mL
    Odor Ether-like
    Sensitivity Light, Air & Moisture Sensitive
    TSCA TSCA
    CAS 100-58-3
    UN Number UN3399
    Health Hazard 2 GHS H Statement
    H224-H261-H314-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    Color Yellow to brown
    Quantity 0.5 mole
    Concentration 3M in ether
    Chemical Name or Material Phenylmagnesium bromide

    GHS H Statement
    H224-H261-H314-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off i

    • Packaged under Argon

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit