Alfa Aesar™ Phenylmercury acetate, 98%

Manufacturer: Alfa Aesar™ 3712518

Quantity 50g

 View more versions of this product

Catalog No. AA3712518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62-38-4
    Linear Formula C6H5HgOCOCH3
    Formula Weight 336.74
    MDL Number MFCD00008691
    UN Number UN1674
    Beilstein 3662930
    Merck Index 14,7300
    Synonym Acetoxy phenylmercury; Acetoxy mercuribenzene
    Chemical Name or Material Phenylmercury acetate

    Specifications

    Formula Weight 336.74
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 146° to 153°C
    Physical Form Powder
    Linear Formula C6H5HgOCOCH3
    CAS 62-38-4
    UN Number UN1674
    Beilstein 3662930
    Health Hazard 2 GHS H Statement
    H300-H372-H314-H318
    Fatal if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    TSCA TSCA
    MDL Number MFCD00008691
    Density 2.4g/mL
    Merck Index 14,7300
    Solubility Information Slightly soluble in water; Soluble in alcohol, benzene and glacial acetic acid
    Odor Odorless
    Quantity 50g
    Synonym Acetoxy phenylmercury; Acetoxy mercuribenzene
    Health Hazard 1 Danger
    Color White to yellow
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENYLMERCURIC ACETATE
    RTECS Number OV6475000
    Chemical Name or Material Phenylmercury acetate
    Assay 98%

    GHS H Statement
    H300-H372-H314-H318
    Fatal if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit