Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Phenylmercury(II) chloride, 96%, Hg 63.5%
Click to view available options
Quantity:
10g
50g
Specifications
Concentration or Composition (by Analyte or Components) | Hg 63.5% |
Quantity | 50g |
Color | White |
Melting Point | 246°C to 250°C |
UN Number | UN2026 |
Merck Index | 14,7301 |
Solubility Information | Soluble in benzene, ether, pyridine; Very slightly soluble in water |
Formula Weight | 313.18 |
Physical Form | Powder |
Percent Purity | 96% |
Show More |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLMERCURIC COMPOUNDS, N.O.S.
EINECSNumber : 202-865-1
RTECSNumber : OW1400000
TSCA : No
Recommended Storage : Ambient temperatures