Alfa Aesar™ Phenylphosphine

Manufacturer: Alfa Aesar™ 3015503

Quantity 1g

 View more versions of this product

Catalog No. AA3015503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 638-21-1
    Formula Weight 110.1
    MDL Number MFCD00002094
    UN Number UN2845
    Chemical Name or Material Phenylphosphine

    Specifications

    Formula Weight 110.1
    Health Hazard 3 GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Health Hazard 2 GHS H Statement
    H250-H252-H332-H315-H319-H335-H227
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    CAS 638-21-1
    Health Hazard 1 Danger
    Density 1.00g/mL
    Odor Stench
    Sensitivity Air & Moisture Sensitive
    Quantity 1g
    Physical Form Liquid
    MDL Number MFCD00002094
    UN Number UN2845
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Boiling Point 160°C
    Chemical Name or Material Phenylphosphine

    GHS H Statement
    H250-H252-H332-H315-H319-H335-H227
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit