Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phenylphosphinic acid, 97%

Manufacturer:  Alfa Aesar™ 30098-18

Quantity 50g

 View more versions of this product

Catalog No. AA3009818

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1779-48-2
MFCD00002131
Benzenephosphinic acid
142.09
UN3261
Phenylphosphinic acid

Specifications

83° to 85°C
1779-48-2
142.09
UN3261
Phenylphosphinic acid
50g
Crystalline
MFCD00002131
Benzenephosphinic acid
97%

Safety and Handling

GHS H Statement
H314-H318-H302

GHS P Statement
P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P310-P301+P312-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit