Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylphosphonic dichloride, 97%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
824-72-6
C6H5Cl2OP
194.98
MFCD00002070
IBDMRHDXAQZJAP-UHFFFAOYSA-N
phenylphosphonic dichloride,phosphonic dichloride, phenyl,benzenephosphonic dichloride,phenylphosphonyl dichloride,dichlorophenylphosphine oxide,phenyldichlorophosphine oxide,benzenephosphonyl chloride,phenylphosphonic acid dichloride,phenylphosphonodichloridic acid,phosphonic dichloride, p-phenyl
69990
dichlorophosphorylbenzene
C1=CC=C(C=C1)P(=O)(Cl)Cl
824-72-6
C6H5Cl2OP
194.98
MFCD00002070
IBDMRHDXAQZJAP-UHFFFAOYSA-N
phenylphosphonic dichloride,phosphonic dichloride, phenyl,benzenephosphonic dichloride,phenylphosphonyl dichloride,dichlorophenylphosphine oxide,phenyldichlorophosphine oxide,benzenephosphonyl chloride,phenylphosphonic acid dichloride,phenylphosphonodichloridic acid,phosphonic dichloride, p-phenyl
69990
dichlorophosphorylbenzene
C1=CC=C(C=C1)P(=O)(Cl)Cl
Specific Gravity 1.39
Flash Point 204°C
Quantity 250mL
Color Yellow to Brown
Density 1.3900g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5570 to 1.5610
Melting Point 3.0°C
Boiling Point 258.0°C
Assay Percent Range 96% min. (Argentometry)
Linear Formula C6H5P(O)Cl2
Beilstein 16, 804
Fieser 01,846; 16,284
Solubility Information Solubility in water: reacts
Formula Weight 194.98
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Phenylphosphonic dichloride, 97%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 212-534-3

TSCA : TSCA