Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylpropiolic Acid 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
637-44-5
C9H6O2
146.15
MFCD00004361
XNERWVPQCYSMLC-UHFFFAOYSA-N
phenylpropiolic acid,3-phenylpropiolic acid,phenylpropynoic acid,3-phenylpropynoic acid,2-propynoic acid, 3-phenyl,phenylacetylenecarboxylic acid,3-phenyl-2-propynoic acid,phenylacetylene monocarboxylic acid,propiolic acid, 3-phenyl,phenyl propiolic acid
69475
3-phenylprop-2-ynoic acid
C1=CC=C(C=C1)C#CC(=O)O
637-44-5
C9H6O2
146.15
MFCD00004361
XNERWVPQCYSMLC-UHFFFAOYSA-N
phenylpropiolic acid,3-phenylpropiolic acid,phenylpropynoic acid,3-phenylpropynoic acid,2-propynoic acid, 3-phenyl,phenylacetylenecarboxylic acid,3-phenyl-2-propynoic acid,phenylacetylene monocarboxylic acid,propiolic acid, 3-phenyl,phenyl propiolic acid
69475
3-phenylprop-2-ynoic acid
C1=CC=C(C=C1)C#CC(=O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color White to Beige
Melting Point 136.0°C to 139.0°C
Assay Percent Range 97%
Linear Formula C6H5C≡CCO2H
Beilstein 09, 633
Solubility Information Solubility in water: freely soluble
Formula Weight 146.15
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material Phenylpropiolic acid, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-285-8

TSCA : TSCA