Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylpyruvic acid, sodium salt monohydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
122049-54-1
C9H7NaO3·H2O
204.16
MFCD00149109
ABWVLWHPVBHFLQ-UHFFFAOYSA-M
sodium phenylpyruvate monohydrate,sodium 2-oxo-3-phenylpropanoate hydrate,phenylpyruvic acid sodium salt monohydrate,phenylpyruvic acid, sodium salt,sodium hydrate ?-phenylpyruvate,l-phenyl pyruvic acid sodium salt,beta-phenylpyruvic acid sodium salt,acmc-2099lh,phenylpyruvic acid sodium hydrate,beta-phenylpyruvic acid, sodium salt
23666336
sodium;2-oxo-3-phenylpropanoate;hydrate
C1=CC=C(C=C1)CC(=O)C(=O)[O-].O.[Na+]
122049-54-1
C9H7NaO3·H2O
204.16
MFCD00149109
ABWVLWHPVBHFLQ-UHFFFAOYSA-M
sodium phenylpyruvate monohydrate,sodium 2-oxo-3-phenylpropanoate hydrate,phenylpyruvic acid sodium salt monohydrate,phenylpyruvic acid, sodium salt,sodium hydrate ?-phenylpyruvate,l-phenyl pyruvic acid sodium salt,beta-phenylpyruvic acid sodium salt,acmc-2099lh,phenylpyruvic acid sodium hydrate,beta-phenylpyruvic acid, sodium salt
23666336
sodium;2-oxo-3-phenylpropanoate;hydrate
C1=CC=C(C=C1)CC(=O)C(=O)[O-].O.[Na+]
Packaging Glass bottle
Quantity 5g
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula C6H5CH2COCO2Na·H2O
Beilstein 10, I, 325
Solubility Information Solubility in water: soluble in water
Formula Weight 204.16
Physical Form Fine Powder
Percent Purity 98%
Chemical Name or Material Phenylpyruvic acid, sodium salt monohydrate, 98%
Show More Show Less

EINECSNumber : 204-053-2