Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phenylselenol

Manufacturer:  Alfa Aesar™ 1572006

Quantity 5g

 View more versions of this product

Catalog No. AA1572006

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

645-96-5
MFCD00000001
Benzeneselenol; Selenophenol
157.03
UN3440
Phenylselenol

Specifications

70° (158°F)
5g
Liquid
MFCD00000001
Benzeneselenol; Selenophenol
71° to 72°C (18mmHg)
645-96-5
157.03
UN3440
Phenylselenol

Safety and Handling

GHS H Statement
H301-H331-H373-H227
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.

GHS P Statement
P210-P260-P261-P301+P310-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, LIQUID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit